    Sort by:
  • Search Within Results
5-Bromo-2-thienylzinc bromide
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   5-Bromo-2-thienylzinc bromide, 0.5M in THF, CAS number: 151073-94-8, Molecular Formula: C4H2Br2SZn, Color: yellow to brown to black, Form: liquid, packaged under Argon in resealable ChemSeal bottles, Size: 50ML