You Searched For: cyclohexylzinc bromide
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (AAH58852-AD)
Supplier: Thermo Scientific Chemicals
Description: Cyclohexylzinc bromide, 0.5M in THF, CAS number: 7565-57-3, Molecular Formula: C6H11BrZn, Color: yellow to brown to black, Form: liquid, packaged under Argon in resealable ChemSeal bottles, Size: 50ML