    Sort by:
  • Search Within Results
4-chlorophenylzinc iodide
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Catalog Number: (AAH58740-AD)
Supplier:  Alfa Aesar
Description:   4-Chlorophenylzinc iodide, 0.5M in THF, CAS number: 151073-70-0, Molecular Formula: C6H4ClIZn, Color: yellow to brown to black, Form: liquid, packaged under Argon in resealable ChemSeal bottles, Size: 50ML