    Sort by:
  • Search Within Results
You Searched for: "phenethylzinc-bromide-solution"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Catalog Number: (AAH58154-AD)
Supplier:  Alfa Aesar
Description:   2-Phenylethylzinc bromide, 0.5M in THF, CAS number: 308796-14-7, Molecular Formula: C8H9BrZn, Color: yellow to brown to black, Form: liquid, Synonym: Phenethylzinc bromide, packaged under Argon in resealable ChemSeal bottles, Size: 50ML