    Sort by:
  • Search Within Results
You Searched for: "4-ethoxy-4-oxobutylzinc-bromide"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   3-(Ethoxycarbonyl)propylzinc bromide, 0.5M in THF, CAS number: 131379-39-0, Molecular Formula: C6H11BrO2Zn, Color: yellow to brown to black, Form: liquid, Synonym: 4-Ethoxy-4-oxobutylzinc bromide, packaged under Argon in resealable ChemSeal bottles, Size: 50ML