You Searched For: Diaion® WA21J
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (AA46418-A1)
Supplier: Thermo Scientific Chemicals
Description: Diaion* WA21J, ion exchange resin, weakly basic porous type, 2.0 meq/ml on poly(styrene-divinylbenzene), Cas Number: 65945-33-7, Color: Pale yellow, Form: 300-1180um Beads, Size: 1KG