    Sort by:
  • Search Within Results
n-3-2-4-4-acetylpiperazin-1-yl-2-methoxyphenyl-amino-5-trifluoromethyl-pyrimidin-4-yl-amino-phenyl-acrylamide
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  BioVision
Description:   CO-1686 (Rociletinib), potent EGFR inhibitor, selectively targeting EGFRL858R/T790M mutant with a Ki value of 21 nM over Wt EGFR (Ki = 303 nM), Purity: ≥98% by HPLC, CAS Number: 1374640-70-6, Molecular Formula: C27H28F3N7O3, Synonyms: AVL-301, Storage: -20 Degree C, Size: 25mg