    Sort by:
  • Search Within Results
You Searched for: "4-ethoxycarbonyl-phenylzinc-bromide"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   4-(Ethoxycarbonyl)phenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal^t bottles