You Searched For: Clopamide
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (89156-938)
Supplier: Enzo Life Sciences
Description: Clinically useful diuretic. Clopamide selectively inhibits the chloride the sodium chloride cotransporter.