    Sort by:
  • Search Within Results
You Searched for: "thionitrobenzoate"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  BioVision
Description:   TNB, A useful DTNB standard, TNB is a useful standard when using DTNB (Ellman’s reagent) for the quantification of thiol (-SH) groups in a sample, Alternate Name/Synonyms: 2-Nitro-5-thiobenzoic acid, CAS Number: 15139-21-6, Purity: ≥98% by HPLC, Molecular Formula: C7H5NO4S, 250 mg