    Sort by:
  • Search Within Results
Fluorescent brightener 28
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  MP Biomedicals
Description:   Soluble in water (80 mg/mL), ethanol (2 mg/mL), acetone, chloroform. Slightly soluble in methanol. Freely soluble in methyethyl ketone, benzene, or et...