    Sort by:
  • Search Within Results
You Searched for: "hydroxypropyl-cyclodextrin"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  MP Biomedicals
Description:   Soluble in water (50 g/100 mL). Solutions may be obtained by stirring 30 minutes at room temperature. Alternatively, sonication with cooling may be em...