Anti-HBsAg Rabbit Polyclonal Antibody (Biotin) Conjugated

Supplier: Fitzgerald Industries
60C-CR2100RB
102013-588EA 553.85 USD
102013-588
Anti-HBsAg Rabbit Polyclonal Antibody (Biotin) Conjugated
Antibodies
HBsAg is the surface antigen of the Hepatitis-B-Virus (HBV). It indicates current Hepatitis B infection.

  • Store at 4 °C for short term storage
  • Aliquot and store at -20 °C for long term storage
  • Avoid repeated freeze/thaw cycles

Type: Tag
Antigen: HBsAg
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hepatitis B Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR