Would you like to log in again?
Bonded AVCS Caps with Septa, 9 mm, National Scientific
Supplier: Thermo Scientific
C5000-62A C5000-62R C5000-62B C5000-62G C5000-72G C5000-62Y C5000-72Y C5000-72B C5000-72R C5000-72C
AVCS (Advanced Vial Closure System) Caps significantly reduce the error rate of analysis that results from evaporation of solvent in vials which are not sealed correctly
  • Re-analysis costs are reduced, sample data integrity increases, and sample throughput increases
  • AVCS closures seal as well as crimp top vials
  • AVCS design allows for consistent sealing, eliminating user-to-user variance
ORDER
Expand All / Collapse All
Description Color Septa Type Supplier No. VWR Catalog Number Unit Price Quantity
Bonded AVCS Caps Black PTFE/Silicone C5000-62A 10222-282 Pack of 100 Retrieving
Bonded AVCS Caps Blue PTFE/Silicone C5000-62B 10222-284 Pack of 100 Retrieving
Bonded AVCS Caps Green PTFE/Silicone C5000-62G 10222-286 Pack of 100 Retrieving
Bonded AVCS Caps Red PTFE/Silicone C5000-62R 10222-290 Pack of 100 Retrieving
Bonded AVCS Caps Yellow PTFE/Silicone C5000-62Y 10222-292 Pack of 100 Retrieving
Bonded AVCS Pre-Slit Caps Black PTFE/Silicone C5000-72B 10222-294 Pack of 100 Retrieving
Bonded AVCS Pre-Slit Caps Green PTFE/Silicone C5000-72G 10222-300 Pack of 100 Retrieving
Bonded AVCS Pre-Slit Caps Grey PTFE/Silicone C5000-72C 10222-298 Pack of 100 Retrieving
Bonded AVCS Pre-Slit Caps Red PTFE/Silicone C5000-72R 10222-306 Pack of 100 Retrieving
Bonded AVCS Pre-Slit Caps Yellow PTFE/Silicone C5000-72Y 10222-308 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty