Hybridization Chamber, Greiner Bio-One
447070
89131-378CS 341.29 USD
89131-378
Hybridization Chamber, Greiner Bio-One
Greiner Bio-One offers a robust and space-saving hybridization chamber.
  • Robust and high-value plastic case
  • Slideholder holds up to 4 slides
  • Easy-to-use
  • Tightly sealed reaction room
  • Temperature-resistant
  • Allows light-sensitive reactions
  • Protects your samples from drying out
  • Polycarbonate and TPR silicone construction
The Hybridization Chamber is an easy-to-handle and cost-saving solution to incubate and store your microarrays, slides, or well plates
The chamber provides a constant humid environment for a wide range of applications and the tightly sealed reaction room prevents the samples from drying out during incubation. Every chamber fits up to 3 standard micro well plates and a supplied slide holder allows parallel processing of up to 4 microscope slides (25 x 75 mm).
Its solid case allows highly light-sensitive reactions within a temperature range from -10 to 90°C.