Would you like to log in again?
Fibronectin-Coated Microplate, 384 Well, Greiner Bio-One
Supplier: GREINER BIO-ONE
781926
Fibronectin Microplates are treated with human plasma-sourced fibronectin to promote attachment and spreading of tissue cell cultures.
  • Convenient
  • Improved attachment of cell cultures
Fibronectin microplates are especially useful for the growth of fibroblasts and other mesenchymally derived cells.
Ordering information: Available by special order.
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Fibronectin-Coated Microplate with Lid, Black/Clear 781926 89093-214 Case of 20 Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty