SP Wilmad-LabGlass Bidwell-Sterling Moisture Traps, SP Industries
Supplier: SP Industries
LG-9095-100
14210-350EA 381.72 USD
14210-350
SP Wilmad-LabGlass Bidwell-Sterling Moisture Traps, SP Industries
Distillation Components Distillation Receivers
Made of borosilicate glass, these Bidwell-Sterling moisture traps are calibrated "to contain" (TC) at 20°C and are used for determining moisture in samples by distilling with an immiscible liquid
Traps consist of vapor tube, return tube, round bottom graduated tube, and Standard Taper 24/40 inner and outer joints.
Certifications: Conforms with ASTM Specification E 123. Also conforms with many methods of American Oil Chemists Society. Reference No. ASTM E 123.