SP Wilmad-LabGlass Ammonia Dissociation Buret, SP Industries
Supplier: SP Industries
LG-4150-100
14007-882EA 1098.68 USD
14007-882
SP Wilmad-LabGlass Ammonia Dissociation Buret, SP Industries
Burets
Graduated from 10 to 90% in 1% subdivisions, this borosilicate glass buret is used to determine the degree of dissociation of ammonia during the nitriding of steel alloys
Buret is 100mL and reads the amount of ammonia absorbed. Stopcocks are glass and a Standard Taper 14/20 joint connects the filling funnel with the buret.