    Sort by:
  • Search Within Results
H-Glu(ANILIDE)-OH
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Bachem Americas
Description:   Sequence: H-Glu(anilide)-OH CAS-No.: 5963-60-0 Mol.weight: 222.24 SumFormula: C₁₁H₁₄N₂O₃ Bulk item no.: E-1935 Synonym(s): H-Gln(Ph)-OH EINECS no.: - ...