Difco™ Minimal Broth Davis, BD
275610 275610
90003-962EA 177.83 USD
90003-962 CA90003-962
Difco™ Minimal Broth Davis, BD
Davis Minimal Broth is recommended for the isolation and characterization of nutritional mutans of Escherichia coli.
  • Citrate and phosphates act as buffers
  • Ammonium sulfate is a nitrogen source
  • Magnesium is a cofactor for many metabolic reactions
  • Dextrose is a carbohydrate energy source
After the mutants are isolated, they are characterized biochemically by growth in minimal broth supplemented with specific growth factors or groups of growth factors.