Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chicken F(ab')2 IgG/IgY Isotype Control (Rhodamine)

Supplier: Rockland Immunochemical
Produced through a multi-stage process that includes delipidation, salt fractionation, ion-exchange chromatography, gel filtration, and affinity chromatography. No contaminating proteins are observed when assayed at a protein concentration of 20mg/mL against anti-whole serum or anti-fragment specific antisera. All immunoglobulin fragments are prepared from highly purified, whole molecules subject to enzymatic digestion.

Ordering information: Purified proteins or fragments are supplied lyophilized. Unconjugated Fc and IgM are supplied as liquids. Conjugated purified proteins or fragments are lyophilized products containing preservative and stabilizer. For additional purified proteins and conjugates, contact your VWR sales representative.

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
IgG/IgY F(ab')2 Fragment 1 mg 003-0004 RL003-0004 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Conjugation Rhodamine
Antibody Type Control
Host Chicken
Isotype IgG/IgY
Antibody Fragment F(ab')2
Protein IgG/IgY F(ab')2 Fragment
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty