TCIAL0267

Supplier: TCI America
L0267-1G
TCL0267-1GEA 78 USD
TCL0267-1G
TCIAL0267
CAS Number: 132843-44-8
MDL Number: MFCD22374096
Molecular Formula: C4F10LiNO4S2
Molecular Weight: 387.09
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 328

Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Bis(pentafluoroethanesulfonyl)imide Lithium Salt
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR