Digital Fluorescence Imaging Analyzer, Full Automated, NanoEnTek USA, INC
JS1000S
75794-852EA 86149.87 USD
75794-852
Digital Fluorescence Imaging Analyzer, Full Automated, NanoEnTek USA, INC
Compact, Compatible, automated fluorescence cell imaging analyzer of live cell imaging for inside your cell culture incubator.
  • Automated imaging system
  • Sensitivity, expandability
  • Networking capability
  • Versatile, powerful S/W
  • Affordability, Easy of use
  • Networking capability
Fully Automated X-Y-Z stage. Multi-channel fluorscence imaging. User-changeable objective lens. Data management with All-in-one PC. Stem cell development. Cell differentiation. Neurite growth.
3 fluorescence;
DAPI: Excitation 390/40, Emission 452/45;
GFP: Excitation 466/40, Emission 525/50;
RFP: Excitation 525/50, Emission 580LP
Accessories information: 4 X, 10 X, 20 X (Olympus) Lens.