Shell Vials for Waters WISP 48-Position, SUN-SRi
Supplier: SUN SRi
200 498 500 160
66008-168PK 65.34 USD
66008-168 66008-082
Shell Vials for Waters WISP 48-Position, SUN-SRi
Vials
15×45 mm vials are available in Clear, Type 1 Class A or Amber, Type I Class B borosilicate glass or high quality polypropylene.
The tear-away design of the polyethylene plug closure ensures excellent penetration and ventilation during sampling. Shell vials are kitted unassembled with the appropriate closures.