Direct Reading Benchtop Flow Meter, Gilmont®, Cole Parmer
Supplier: Cole Parmer
323012
29895-050EA 293.79 USD
29895-050
Direct Reading Benchtop Flow Meter, Gilmont®, Cole Parmer
Unshielded flowmeter is suitable for low-pressure applications
  • Modular components allow for meter flexibility across a wide range of flow rates
  • Multiple end connections to suit process requirements
Connect the flowtube end directly to tubing having the proper inner diameter. Glass taper joints are an alternate connection option which slip on the PTFE stops that contain the float.