Anti-p70 S6 Kinase Rabbit Polyclonal Antibody

Supplier: PromoCell
PK-AB577-3485
10174-378EA 520.24 USD
10174-378
Anti-p70 S6 Kinase Rabbit Polyclonal Antibody
Antibodies
Rabbit Anti-Human/Mouse/Rat p70 S6 Kinase

Type: Primary
Antigen: p70 S6 Kinase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR