Anti-Phospho-JKK/SEK1/MKK4 Rabbit Polyclonal Antibody

Supplier: PromoCell
PK-AB577-3478
10174-370EA 598.29 USD
10174-370
Anti-Phospho-JKK/SEK1/MKK4 Rabbit Polyclonal Antibody
Antibodies
Rabbit Anti-Human/Mouse/Rat Phospho-JKK/SEK1/MKK4

Type: Primary
Antigen: Phospho-JKK/SEK1/MKK4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: The antibody recognizes phosphorylated SEK1/MKK4/JKK1 of human, mouse, and rat origins.
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR