Anti-Phospho-Stat2 Rabbit Polyclonal Antibody

Supplier: PromoCell
PK-AB577-3469
10174-346EA 596.67 USD
10174-346
Anti-Phospho-Stat2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Anti-Human/Mouse Phospho-Stat2

Type: Primary
Antigen: Phospho-Stat2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: The antibody recognizes ~113 kDa phosphorylated Stat2 (Tyr689) of human and mouse origins. Reactivity to other species has not been tested.
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR