Anti-FAK Rabbit Polyclonal Antibody

Supplier: PromoCell
PK-AB577-3399
10175-572EA 557.41 USD
10175-572
Anti-FAK Rabbit Polyclonal Antibody
Antibodies
Rabbit Anti-Human/Mouse/Rat FAK

Type: Primary
Antigen: FAK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR