Anti-Cathepsin G Rabbit Polyclonal Antibody

Supplier: PromoCell
PK-AB577-3370
10174-248EA 557.41 USD
10174-248
Anti-Cathepsin G Rabbit Polyclonal Antibody
Antibodies
Rabbit Anti-Human Cathepsin G

Type: Primary
Antigen: Cathepsin G
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR