Anti-Annexin V Rabbit Polyclonal Antibody

Supplier: PromoCell
PK-AB577-3357
10174-210EA 596.67 USD
10174-210
Anti-Annexin V Rabbit Polyclonal Antibody
Antibodies
Rabbit Anti-Human/Mouse/Rat Annexin V

Type: Primary
Antigen: Annexin V
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: The antibody detects Annexin V from samples of human, mouse, and rat origin.
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR