Anti-IkB-alpha Rabbit Polyclonal Antibody

Supplier: PromoCell
PK-AB577-3252
10174-150EA 543.78 USD
10174-150
Anti-IkB-alpha Rabbit Polyclonal Antibody
Antibodies
IkB-alpha

Type: Primary
Antigen: IkB-alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR