Anti-PKA Rabbit Polyclonal Antibody

Supplier: PromoCell
PK-AB577-3115
10174-072EA 543.78 USD
10174-072
Anti-PKA Rabbit Polyclonal Antibody
Antibodies
Rabbit Anti-Human/Mouse/Rat/Bovine Protein Kinase A

Type: Primary
Antigen: PKA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR