HiVeg™ Cetrimide Agar Base, HiMedia Laboratories
Supplier: HiMedia Laboratories
MV024-500G
61000-262EA 95.05 USD
61000-262
HiVeg™ Cetrimide Agar Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation of Pseudomonas aeruginosa from clinical specimens
  • Acts as a quaternary ammonium compound
  • Determines ability of organism to produce fluorescein and pyocyanin
  • Causes nitrogen and phosphorus to be released from bacterial cells
It acts as a quaternary ammonium compound, cationic detergent that causes nitrogen and phosphorus to be released from bacterial cells.
Also used for determining the ability of an organism to produce fluorescein and pyocyanin.