2,2,3,3,3-Pentafluoro-1-propanol derivatisation reagent

Supplier: RESTEK CORP
Warning

Synonyms: 1H,1H-Pentafluoro-1-propanol1H,1H-Pentafluoropropanol-1PFPOH (pentafluoropropanol)

35625
75835-422EA 117.77 USD
75835-422
2,2,3,3,3-Pentafluoro-1-propanol derivatisation reagent
2,2,3,3,3-Pentafluoro-1-propanol

Derivatization reagent by acylation

Formula: C₃H₃F₅O
MW: 150.05 g/mol
MDL Number: MFCD00004673
CAS Number: 422-05-9
EINECS: 207-012-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR