MMP-2/MMP-9 Inhibitor I, Calbiochem®

Supplier: MilliporeSigma

444241-5MG
80502-986EA 304.77 USD
80502-986
MMP-2/MMP-9 Inhibitor I, Calbiochem®
MMP-2/MMP-9 Inhibitor I

MMP-2/MMP-9 Inhibitor I controls the biological activity of MMP-2/MMP-9


  • Orally active in animal models of tumor growth and metastasis
  • IC50=310 nM and 240 nM against MMP-2 and MMP-9


This small molecule/inhibitor is primarily used for protease inhibitors applications.

Formula: C₂₁H₁₉NO₄S
MW: 381.4 g/mol
CAS Number: 193807-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR