    Sort by:
  • Search Within Results
Ferroceneboronic acid
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  TCI America
Description:   [Cyclic boronating reagent for GC/MS] CAS Number: 12152-94-2 MDL Number: MFCD00059074 Molecular Formula: C10H11BFeO2 Molecular Weight: 229.85 Pur...