    Sort by:
  • Search Within Results
cis-3-Aminocyclohexanecarboxylic acid
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  TCI America
Description:   CAS Number: 16636-51-4 MDL Number: MFCD09753647 Molecular Formula: C7H13NO2 Molecular Weight: 143.19 Purity/Analysis Method: 96.0% (GC,T) For...