Chlorophenol red sodium salt
Supplier:
Alfa Aesar
Description:
Transition interval: pH 4.8 (yellow) to 6.4 (red)
Supplier:
TCI America
Description:
CAS Number: 123333-64-2
MDL Number: MFCD00151199
Molecular Formula: C19H12Cl2O5S
Molecular Weight: 445.24
Form: Crystal
Color: Deep Yellow Red
Catalog Number:
(AAAL10364-06)
Supplier:
Alfa Aesar
Description:
Transition interval: pH 4.8 (yellow) to 6.4 (red)
Supplier:
BeanTown Chemical
Description:
CAS: 123333-64-2; MDL No: MFCD00151199
Liquid; Molecular Formula: C19H11Cl2NaO5S; MW: 445.26
Density (g/mL): 0.99
Supplier:
Acros Organics
Description:
Chlorophenol Red, sodium salt, pure, water soluble, Cas Number: 123333-64-2, Purity Grade: Pure, Molecular Formula: C19H11Cl2NaO5S, Formula Weight: 445.25, Synonym: 3', 3''-Dichlorophenolsulfonephthalein, sodium salt, Size: 50G
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the
![]()
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the
![]()
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
* Please note if your account is within the State of California two of these pieces of identification will be required.
???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
|
|||||||||