    Sort by:
  • Search Within Results
9-9-bis-3-amino-4-hydroxyphenyl-fluorene
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  TCI America
Description:   CAS Number: 20638-07-7 Molecular Formula: C25H20N2O2 Molecular Weight: 380.45 Purity/Analysis Method: 98.0% (HPLC) Form: Crystal Flash Point (...