You Searched For: 5-Carboxyfluorescein


10  results were found

SearchResultCount:"10"

Sort Results

List View Easy View

Rate These Search Results

Catalog Number: (TCC2477-100MG)
Supplier: TCI America
Description: CAS Number: 76823-03-5 MDL Number: MFCD00036874 Molecular Formula: C21H12O7 Molecular Weight: 376.32 Purity/Analysis Method: 97.0% (HPLC,T) F...

Supplier: Thermo Scientific Chemicals
Description: 5-CARBOXYFLUORESCEIN 95% 100MG

Supplier: Thermo Scientific Chemicals
Description: 5-Carboxyfluorescein, single isomer, Purity: 97%, CAS number: 76823-03-5, Molecular Formula: C21H12O7, Color: Orange, Color: Orange, Size: 250mg
Catalog Number: (89165-966)
Supplier: Enzo Life Sciences
Description: Amine reactive dye


Supplier: Bachem Americas
Description: Sequence: 5-Carboxy-fluorescein
Synonym(s): 5-FAM#4-(6-Hydroxy-3-oxo-3H-xanthen-9-yl)-isophthalic acid

Supplier: Ambeed
Description: 5-FAM 1G

New Product

Supplier: AAT BIOQUEST INC
Description: 5-FAM is the purified single isomer of carboxyfluorescein.

Minority or Woman-Owned Business Enterprise Small Business Enterprise

Catalog Number: (89139-260)
Supplier: Biotium
Description: 5-Carboxyfluorescein has a pKa of 6.5 and can be used as a pH sensor. The excitation spectrum of the dye responds to pH similarly to that of BCECF. Ca...


Supplier: Anaspec Inc
Description: 5-FAM is a single isomer. It is one of the most popular green fluorescent reagents used for in situ labeling peptides, proteins and nucleotides. It ha...

Supplier: Adipogen
Description: The amine-reactive fluorescein derivatives are probably the most common fluorescent derivatization reagents for covalently labeling proteins. Carboxyf...

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 10 of 10
no targeter for Bottom