Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
You Searched for: "n-trimethylsilyl-imidazole"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  TCI America
Description:   TMS-PZ kit is a trimethylsilylating reagent, for wet or dry sugars, mono- or poly-saccharides.