Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
You Searched for: "2-4-diamino-6-2-2-methyl-1-imidazolyl-ethyl-1-3-5-triazine"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  TCI America
Description:   CAS Number: 38668-46-1 Molecular Formula: C9H13N7 Molecular Weight: 219.25 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point ...