You Searched For: [Di-tert-butyl(chloro)phosphine]palladium(II) dichloride dimer
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (TCD3704-100MG)
Supplier: TCI America
Description: CAS Number: 386706-33-8 MDL Number: MFCD05664440 Molecular Formula: C16H36Cl6P2Pd2 Molecular Weight: 715.95 Purity/Analysis Method: 97.0% (T)...