    Sort by:
  • Search Within Results
(S)-(+)-2,2,2-Trifluoro-1-(9-anthryl)ethanol
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  TCI America
Description:   [e.e. Determination Reagent by NMR] CAS Number: 60646-30-2 MDL Number: MFCD00062968 Molecular Formula: C16H11F3O Molecular Weight: 276.26 Purity/...