    Sort by:
  • Search Within Results
(2R,3R,5S,6S)-5,6-Bis(hydroxymethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  TCI America
Description:   CAS Number: 173371-55-6 Molecular Formula: C10H20O6 Molecular Weight: 236.26 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (...