    Sort by:
  • Search Within Results
(1R,2S)-(-)-trans-2-Phenyl-1-cyclohexanol
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  TCI America
Description:   CAS Number: 98919-68-7 MDL Number: MFCD00075488 Molecular Formula: C12H16O Molecular Weight: 176.26 Purity/Analysis Method: 98.0% (GC) Form: ...