Anti-CD206 Mouse Monoclonal Antibody (IgG1) [clone: 15-2]

Supplier: Bio-Rad
MCA2155T
102691-772EA 147.69 USD
102691-772
Anti-CD206 Mouse Monoclonal Antibody (IgG1) [clone: 15-2]
Antibodies
Mouse anti Human CD206 monoclonal antibody, clone 15-2 recognizes human macrophage mannose receptor 1, also known as CD206 or C-type lectin domain family 13 member D-like. CD206 is a ~175 kDa single pass type I transmembrane glycoprotein belonging to the group of pattern recognition receptors.

CD206 has multiple carbohydrate recognition motifs
It act as receptor for bacteria, fungi and other pathogens
CD206 is predominantly expressed in tissue macrophages and dendritic cells

Type: Primary
Antigen: CD206
Clonality: Monoclonal
Clone: 15-2
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR