Heterotrophic Agar, Plates, Northeast Lab Services

Supplier: Northeast Lab Services
P1320
101373-988PK 98.71 USD
101373-988
Heterotrophic Agar, Plates, Northeast Lab Services
Media for Microbiology Prepared Media for Microbiology
This Heterotrophic Agar is contained in a standard Petri Dish.

  • Standard Petri Dish
  • Fill Volume: 20 mL
  • Prepared Medium
  • Size: 15 mm x 100 mm

The petri dish is1 5 mm x 100 mm, and the fill volume is 20 mL.

NEL's plated media operation utilizes carefully selected and validated ingredients for contaminant-free products with a high degree of consistency from batch to batch.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR