NEMA 4X Heavy Duty Rugged Xenon Stroboscopes, Nidec Shimpo
Supplier: Nidec-Shimpo
DT-311J
75836-588EA 1391.68 USD
75836-588
NEMA 4X Heavy Duty Rugged Xenon Stroboscopes, Nidec Shimpo
Where powerful illumination and rugged speed measuring accuracy is required for the textile industry, the DT-311J will perform where other stroboscopes will not.
  • NEMA 4X IP65 housing for extreme durability
  • External triggering control, automatically synchronizes to input signal
  • Long Life 1500FPM flashtubes are designed for extra long life spans
The DT-311J is used to set injection timing, injection pressure, timing of shedding and weft running time of weaving machinery. Units combine rugged NEMA 4X (IP65) housing with high performance accuracy, special phase shifting, with shock absorbing components and top mounted handle, allows the equipment operator maximum flexibility when setting the timing of weaving machinery.